missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Permabond™ SE-52 GC Capillary Column
Brand: Macherey-Nagel™ 723054.25
This item is not returnable.
View return policy
Specifications
0.4 mm | |
0.25 mm | |
G27, G36 | |
300°C to 320°C | |
25 m | |
1 Each |
GC Capillary Column | |
Permabond™ SE-52 | |
Gas Chromatography (GC) | |
0.25 μm | |
Permabond™ SE-52 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction