missing translation for 'onlineSavingsMsg'
Learn More

Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized, Thermo Scientific Chemicals

Oxidation reagent

75.90 € - 252.00 €

Chemical Identifiers

CAS 79-21-0, 7722-84-1, 64-19-7
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10302633
View Documents Promotion Details
Thermo Scientific Acros
257751000
100 mL
75.90 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10670395
View Documents Promotion Details
Thermo Scientific Acros
257755000
500 mL
252.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

79-21-0, 7722-84-1, 64-19-7
peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
ethaneperoxoic acid
KFSLWBXXFJQRDL-UHFFFAOYSA-N
6585
CC(=O)OO

Specifications

79-21-0, 7722-84-1, 64-19-7
1.1300g/mL
62°C
C2H4O3
15,7263
peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
KFSLWBXXFJQRDL-UHFFFAOYSA-N
ethaneperoxoic acid
6585
Peroxyacetic acid
-44.0°C
105.0°C
Plastic bottle
01,785; 02,307; 03,219; 04,372; 05,505; 06,452; 08,386; 11,402; 12,378
1.13
Solubility in water: soluble. Other solubilities: freely soluble in sulfuric acid, freely soluble in alcohol and ether
CC(=O)OO
76.05
76.05
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
Heating may cause a fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep/Store away from clothing/ combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-186-8

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only