missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic acid, 75-80% aq. soln., Thermo Scientific Chemicals

134.67 € - 551.75 €

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341229
View Documents
Thermo Scientific Alfa Aesar
011413.06
5 g
134.67 €
5g
Please to purchase this item. Need a web account? Register with us today!
11351229
View Documents
Thermo Scientific Alfa Aesar
011413.14
25 g
551.75 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Perrhenic acid is used in the manufacturing of superconducting synthetic metals and X-ray targets. It is used as a hydrocracking catalyst in petroleum industry and used for the dehydrogenation process and epoxidation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13768-11-1
251.21
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
83718
O[Re](=O)(=O)=O
HO4Re
MFCD00011326
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
hydroxy(trioxo)rhenium

Specifications

13768-11-1
Liquid
HO4Re
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
hydroxy(trioxo)rhenium
83718
Perrhenic acid: 80.0%; Water: 20.0%
23 hPa (17mm Hg) at 20°C
Perrhenic acid
Colorless through Yellow
75-80%
MFCD00011326
Soluble in water.
O[Re](=O)(=O)=O
251.21
251.26
Colorless to pale yellow liquid
2.42
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: I

missing translation for 'einecsNumber' : 237-380-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Perrhenic acid, 75-80% aqueous solution

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Perrhenic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.