missing translation for 'onlineSavingsMsg'
Learn More

Manutan Personal effects locker, 8 compartment, grey flat top 920mm x 250mm x 160mm

Brand:  Manutan A330446

Encompass_Preferred

Product Code. 14874341

  • 288.95 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Compact storage for items such as mobile phones, wallets or cameras. Four or eight door small compartment steel lockers, can be floor standing or wall mounted. Supplied with a key cam lock (two keys supplied).
  • Eight doors
  • Flat top
  • Grey body/door colour
  • Dimensions (H x W x D) : 920mm x 250mm x 160mm
  • Compartment dimensions (H x W x D) : 75mm x 165mm x 135mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.