missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius PES Tangential Flow Filtration Cassette for up to 0.5 L
Plug-and-play tangential flow filtration cassette that is made from PES, for ultrafiltration and diafiltration of intermediate sample volumes.
232.00 €
Specifications
Filter Type | Ultrafiltration |
---|---|
Format | Cassette |
For Use With (Equipment) | Peristaltic Pump |
Material | Polycarbonate |
Material (Membrane) | PES |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
30178726
|
Sartorius
VF-S050P0005-IV |
5 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178738
|
Sartorius
VF-S050P0010-IV |
10 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178719
|
Sartorius
VF-S050P0030-IV |
30 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178734
|
Sartorius
VF-S050P0050-IV |
50 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178728
|
Sartorius
VF-S050P0100-IV |
100 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178716
|
Sartorius
VF-S050P0300-IV |
300 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30178730
|
Sartorius
VF-S050P1000-IV |
1000 kDa |
232.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: PES
- Recommended feed volume: 100 to 500 mL
Specifications
Ultrafiltration | |
Peristaltic Pump | |
PES | |
Non-sterile | |
0.5 L |
Cassette | |
Polycarbonate | |
0.1 to 0.5 L | |
TFF cassette |