Learn More
Pesticide Mix 3, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 3 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand:  SPEX Certiprep SPXPR-3
Product Code. 16300488
  • 250.00 € / 1mL
Explore more special offers
Description
Description
  • Pesticide Mix 3 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Specifications
Propargite (omite)
Carbaryl
Myclobutanil (systhane)
Dimethomorph
Etoxazole
Spirodiclo
Glass Ampule
97.4
99.9
98.6
98.4
98.9
99.5
99.6
99.5
99.5
98.5
99
96
98.9
99
99.4
350.472 g/mol
201.221 g/mol
288.775 g/mol
387.8567 g/mol
359.410 g/mol
411.319 g/mol
291.715 g/mol
229.159 g/mol
376.488 g/mol
260.377 g/mol
141.001328 g/mol
373.631 g/mol
223.1635 g/mol
320.3647 g/mol
305.334 g/m
100μg/mL
1 mL
C19H26O4S
C12H11NO2
C15H17ClN4
C21H22ClNO4
C21H23F2NO2
C21H24Cl2O4
C8H10ClN5O3S
C9H6F3N3O
C25H28O3
C7H17O2PS3
C2H8NO2PS
C11H15BrClO3PS
C7H14NO5P
C12H17O4PS2
C11H20N3O3PS