Learn More
Pesticide Mix 4, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 4 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand:  SPEX Certiprep SPXPR-4
Product Code. 16310488
  • 204.00 € / 1mL
Explore more special offers
Description
Description
  • Pesticide Mix 4 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Specifications
Specifications
Epn
Dichlorvos
Edifenphos
Ethion
Fenitrothion
Ethyl parathion
Methyl parathion
Glass Ampule
99.1
98.8
97.5
99.5
98
99
99.6
99.5
99.5
99.2
99.2
99.5
98
99.5
98
323.304 g/mol
220.976 g/mol
310.371 g/mol
384.476 g/mol
277.234 g/mol
291.261 g/mol
263.207 g/mol
183.166 g/mol
274.404 g/mol
278.328 g/mol
304.345 g/mol
350.59 g/mol
437.148 g/mol
248.185 g/mol
265.911 g/mol
100μg/mL
1 mL
C14H14NO4PS
C4H7Cl2O4P
C14H15O2PS2
C9H22O4P2S4
C9H12NO5PS
C10H14NO5PS
C8H10NO5PS
C4H10NO3PS
C8H19O2PS3
C10H15O3PS2
C12H21N2O3PS
C9H11Cl3NO3PS
C12H4Cl2F6N4OS
C12H6F2N2O2
C8Cl4N2