Learn More
Pyrex™ Borosilicate Glass Pestle
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
Brand:  Pyrex 718347
19.14 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12682022
  • 20.65 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex™ Mortars (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rough
Borosilicate Glass
125mm
Pestle
Pyrex™ Mortar