missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Pestle
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
Brand: Pyrex 718342
15.30 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ideal for crushing solids into powders for easier handling
- Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion
- Available in a rough or glazed finish
- For use with Pyrex™ Mortars (sold separately)
Specifications
Rough | |
Borosilicate Glass | |
110mm |
Pestle | |
Pyrex™ Mortar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction