missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ PET Liquid Scintillation Vials

For optimal performance in the laboratory. Wheaton™ PET Liquid Scintillation Vials offer low permeability to solvents and minimal background counts.

Brand:  Wheaton™ 986744

478.76 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10517724

  • 503.00 € / Pack of 1000
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With caps attached
  • Available linerless or with foamed polyethylene liner
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Linerless
PET
Polypropylene
61 mm
Scintillation Vial
27 mm
20 mL
Screw Cap
1000 Pk.
22-400
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.