missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PETG Erlenmeyer Flask with Vent Cap
BIOPHARMAGREENER_CHOICE

Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.

Brand:  Corning™ 431282

269.77 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 15873875

  • 317.00 € / Pack of 4
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning™ Erlenmeyer and Fernbach culture flasks are ideal for shaker culture applications and storage. Replacement caps available for 2 L, 3 L, and 5 L flask sizes.

  • Polyethylene terephthalate glycol-modified (PETG) construction: USP Class VI material provides excellent optical clarity and mechanical strength
  • Sizes range from 2 L to 5 L
  • Molded-in graduations for accuracy (±5%)
  • Vent cap for continuous gas exchange while ensuring sterility and preventing leakage
  • Individually packaged
  • Sterility Assurance Level (SAL) of 10−6
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nonpyrogenic, DNase/RNase-free
Polypropylene
4/Cs.
Fernbach
Non-baffled
Orange
3 L
1/Pk.
Sterile
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.