Learn More
3M™ Petrifilm™ E.coli/Coliform Count Plates
Provide a cost effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples.
Brand: 3M™ 7100039397
This item is not returnable.
View return policy
Description
- Confirmed E. coli results in 24-48 hours.
- Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
- Interpretation can be done with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
- This plate has been awarded Official Methods of Analysis OMA #991.14.for Food OMA # 998.08 for Poultry, Meat and Sea Food
- Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
E. coli | |
Petrifilm E.coli/Coliform Count Plates | |
Environmental Test | |
500 Pack |