Learn More
3M™ Petrifilm™ E.coli/Coliform Count Plates
Provide a cost effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples.
Brand:  3M™ 7100039312
Encompass
Product Code. 17213754
  • 109.62 € / Pack of 50
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Confirmed E. coli results in 24-48 hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Interpretation can be done with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
  • This plate has been awarded Official Methods of Analysis OMA #991.14.for Food OMA # 998.08 for Poultry, Meat and Sea Food
  • Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Petrifilm E.coli/Coliform Count Plates
50