missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
51.30 € - 393.00 €
Chemical Identifiers
CAS | 64742-49-0 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.178 |
MDL Number | MFCD00081849 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
PubChem CID | 7892 |
SMILES | CCCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10151720
|
Fisher Chemical
P/1760/15 |
1 L | Amber Glass Bottle |
51.30 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10568060
|
Fisher Chemical
P/1760/17 |
2.5 L | Amber Glass Bottle |
95.20 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11433633
|
Fisher Chemical
P/1760/PB17 |
2.5 L | Plastic-coated Glass Bottle |
104.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10161720
|
Fisher Chemical
P/1760/21 |
5 L | Fluorinated HDPE Bottle |
234.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10204350
|
Fisher Chemical
P/1760/25 |
25 L | Metal Can |
393.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
Specifications
64742-49-0 | |
40°C | |
C6H14 | |
38kPa at 20°C | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C | |
86.178 |
Colorless | |
Liquid | |
MFCD00081849 | |
0.45 mm2/s at 25°C (ASTM D445) | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
2-methylpentane | |
7892 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Petroleum ether, boiling range 40-60°C
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.