Learn More
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
36.28 € - 329.64 €
Chemical Identifiers
CAS | 64742-49-0 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.178 |
MDL Number | MFCD00081849 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
PubChem CID | 7892 |
SMILES | CCCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10702611
|
Thermo Scientific Acros
180210010 |
1 L | Glass Bottle |
36.28 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184000
|
Thermo Scientific Acros
180210025 |
2.5 L | Glass Bottle |
66.43 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10277682
|
Thermo Scientific Acros
180210050 |
5 L | Plastic Drum |
105.53 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10707403
|
Thermo Scientific Acros
180210250 |
25 L | Metal Drum |
329.64 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64742-49-0 | |
86.178 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 |
C6H14 | |
MFCD00081849 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C |
Specifications
64742-49-0 | |
40.0°C to 65.0°C | |
1.3660 to 1.3710 | |
C6H14 | |
0.45 mm2/s (25°C) | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
Petroleum ether | |
7892 | |
Petroleum ether |
0.6500g/mL | |
−45°C | |
0.65 | |
MFCD00081849 | |
14, 5489 | |
Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil | |
CCCC(C)C | |
86.178 | |
ACS Reagent |
Safety and Handling
GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Petroleum ether, boiling range 40-60°C
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.