Learn More
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Petroleum ether |
---|---|
Melting Point | -30.0°C |
CAS | 64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5 |
Density | 0.7400g/mL |
Boiling Point | 100.0°C to 140.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10591291
|
Thermo Scientific Acros
233020025 |
2.5 L | Glass Bottle |
N/A
|
N/A | ||||
10368703
|
Thermo Scientific Acros
233020100 |
10 L | Plastic Drum |
N/A
|
N/A | ||||
10037252
|
Thermo Scientific Acros
233020250 |
25 L | Metal Drum |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Petroleum ether | |
64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5 | |
100.0°C to 140.0°C | |
Authentic | |
1.4030 to 1.4080 | |
0.74 | |
0.76 mm2/s (25°C) | |
Solubility in water: insoluble. | |
Extra Pure |
-30.0°C | |
0.7400g/mL | |
2°C | |
Liquid | |
0.005% max. | |
MFCD00081849 | |
14, 5489 | |
Petroleum ether |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Petroleum ether, boiling range 100-140°C
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Naphtha, petroleum,hydrotreated light,Hexane, Cyclohexane
RUO – Research Use Only