missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss. p.a., high boiling, bp 60-80°C, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
Packaging:
Glass Bottle
Specifications
Specifications
CAS | 101316-46-5 |
Density | 0.660 to 0.680g/mL at 20°C |
Quantity | 4 x 2.5 L |
Flash Point | −40°C (−40°F) |
Packaging | Glass Bottle |
MDL Number | MFCD00081849 |
UN Number | UN1268 |
Grade | puriss. p.a. |
Chemical Name or Material | Petroleum Ether |
Safety and Handling
Product Identifier
- Petroleum ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrocarbons, C6, n-Alkanes, Isoalkanes,Cyclics, n-Hexane rich, Hydrocarbons, C6 - C7, n-Alkanes, I
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'