Learn More
PF-05085727, MedChemExpress
MedChemExpress PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.
Brand: Medchem Express HY-102050
This item is not returnable.
View return policy
Description
PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.Specifications
PF-05085727 | |
Yellow | |
C20H18F3N7 | |
DMSO : 62.5 mg/mL (151.19 mM; Need ultrasonic) | |
413.4 | |
98.0% | |
Solid |
1415637-72-7 | |
Research | |
5 mg | |
CN1C2=NC=NC(N3CCC3)=C2C(C4=C(C5=CC=C(C(F)(F)F)C=C5)N(C)N=C4)=N1 | |
413.4 | |
Research | |
Neuroscience-Neuromodulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PF-05085727, MedChemExpress