missing translation for 'onlineSavingsMsg'
Learn More

PF-05180999, MedChemExpress

MedChemExpress PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.

Brand:  Medchem Express HY-111371

Encompass_Preferred

 View more versions of this product

Product Code. 16427157

  • 112.05 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.
Specifications

Specifications

PF-05180999
Off-White
C19H17F3N8
PF-999
CC1=NC(C2=C(C3=CC=C(C(F)(F)F)C=N3)N(C)N=C2)=C4N1N=CN=C4N5CCC5
414.39
Research
Neuroscience-Neuromodulation
1394033-54-5
Research
5 mg
DMSO : 50 mg/mL (120.66 mM; Need ultrasonic)
414.39
98.93%
Solid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
PF-05180999, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research purposes only