missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-05180999, MedChemExpress
MedChemExpress PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.
Brand: Medchem Express HY-111371

This item is not returnable.
View return policy
Description
PF-05180999 (PF-999) is a phosphodiesterase 2A (PDE2A) inhibitor, with an IC50 of 1.6 nM.Specifications
PF-05180999 | |
Off-White | |
C19H17F3N8 | |
PF-999 | |
CC1=NC(C2=C(C3=CC=C(C(F)(F)F)C=N3)N(C)N=C2)=C4N1N=CN=C4N5CCC5 | |
414.39 | |
Research | |
Neuroscience-Neuromodulation |
1394033-54-5 | |
Research | |
50 mg | |
DMSO : 50 mg/mL (120.66 mM; Need ultrasonic) | |
414.39 | |
98.93% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PF-05180999, MedChemExpress
Research purposes only
Spot an opportunity for improvement?Share a Content Correction