Learn More
Veltek PF-4 Sterile Cleanroom Facemask
Constructed of 3 layers of non-woven material to help prevent skin irritation or allergy. Coated metal nosepiece keeps the mask secure. Manufactured in a clean environment and is lot traceable.
Brand:  Veltek PF-4SM-S-2-2029
Product Code. 16853203
  • 1375.00 € / Pack of 500
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterile facemasks are individually packaged and lot traceable
  • 10.0 to 20.0 kGy gamma irradiated
  • Hypoallergenic, soft surface layers prevent skin irritation and allergy problems
  • Silicon-free headloop OP bands
  • Ultrasonic bonded loop bands ensure a cleaner mask, reducing contamination and outgassing
Specifications
Specifications
Cleanroom Face Mask
Polypropylene, Rayon
Certificate of Irradiation; Certificate of Sterility; Certificate of Conformance; Lot traceable
Five year, closed package expiration date. Individually packaged
94.74% per EN 14683:2019
Yes
500/Pk.
White
Sterile
3-ply
Single-use
Cleanrooms
PF-4