missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution pH 2, Fisher Chemical™
Shop All Reagecon Products

Click to view available options
Quantity:
5 L
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
Color | Undesignated |
pH | 2 |
Quantity | 5 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pH Buffer Solution pH 2, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction