missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution pH 5, Fisher Chemical™
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
10 L
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
Color | Undesignated |
pH | 5 |
Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction