missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution pH 6, Fisher Chemical™
Click to view available options
Quantity:
5 L
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
Color | Undesignated |
pH | 6 |
Quantity | 5 L |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'