Learn More
Hanna Instruments™ pH Edge™ Electrode
Brand:  Hanna Instruments™ HI12300
Product Code. 15188239
  • 419.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
3.5 mm Jack
PEI
-5°C to +70°C
7.5 mm
Ceramic Single, 15 to 20 μL/h
0 to 13
1 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ pH Edge™ Electrode >