Learn More
pH Electrode Storage Solution, Reagecon™
BIOPHARMA
Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.
Brand:  Reagecon ESS5
 View more versions of this product
Product Code. 11533182
  • 110.00 € / 500mL
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The response, accuracy and lifespan of pH electrodes can be substantially improved through correct handling, cleaning and storage. Designed for routine equipment maintenance, these solutions are safe to use with any sensitive electrode. Electrode storage solutions are made from potassium chloride, free from silver ion, and are suitable for all Blueline electrodes.
Specifications
Specifications
Potassium Chloride solution, without Silver ion
pH and Reference Electrodes, All Blueline Electrodes
500 mL
3 mol/L KCl
Electrode Storage