Learn More
pH Electrode Storage Solution, Reagecon™
Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.
Brand: Reagecon ESS01
This item is not returnable.
View return policy
Description
The response, accuracy and lifespan of pH electrodes can be substantially improved through correct handling, cleaning and storage. Designed for routine equipment maintenance, these solutions are safe to use with any sensitive electrode. Electrode storage solutions are made from potassium chloride, free from silver ion, and are suitable for all Blueline electrodes.Specifications
Potassium Chloride solution, without Silver ion | |
pH and Reference Electrodes, All Blueline Electrodes | |
1000 mL |
3 mol/L KCl | |
Electrode Storage |