missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ pH-Fix pH Indicator Strips

Highly appreciated standard strips for many pH testing applications.

16.05 € - 22.90 €

Specifications

Quantity 100 Strips
Type pH Indicator Paper
Length (Metric) 85 mm
Width (Metric) 6 mm
View More Specs

Products 3
Product Code Brand pH Range Price Quantity & Availability  
Product Code Brand pH Range Price Quantity & Availability  
15705802
View Documents
Macherey-Nagel™
92110
0 to 14
19.90 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15324194
View Documents
Macherey-Nagel™
92190
1.7 to 3.8
16.05 €
100 units
In Stock
to see stock.
Add to basket
 
15364184
View Documents
Macherey-Nagel™
92118
2 to 9
22.90 €
100 units
In Stock
to see stock.
Add to basket
 
Description

Description

Highly appreciated standard strips for many pH testing applications.

  • Easy dip and read procedure provides reliable results in seconds
  • Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
  • Dye does not wash out, so clothes stay clean and samples remain pure
Specifications

Specifications

100 Strips
pH Indicator Paper
85 mm
6 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.