Learn More
SI Analytics™ pH Glass Electrode with Screw Plug Head
SI Analytics™ pH Glass Electrode with Screw Plug Head is constructed of a glass shaft with an Ag/AgCl reference system.
Brand:  SI Analytics™ 285111483
Product Code. 10450582
  • 278.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • pH glass electrodes
  • With screw plug head
  • Glass shaft
  • Ag/AgCl-reference system
  • Sphere membrane
  • S-glass
  • Zeropoint pH = 7.0
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Plug Head
2 to 14
10°C to 135°C
12 mm
S-Glass
Sphere
120 mm