missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pH Gradient Buffer Concentrates

Product Code. 15105634
missing translation for 'orderingAttributeHoverText'
Description:
CX-1 pH Gradient Buffer A
CX-1 pH Gradient Buffer B
CX-1 pH Gradient Buffer Kit, Buffer A + Buffer B
CX-1 pH Gradient High Resolution Kit, Buffer A + Buffer B
CX-1 pH Gradient High Throughput Kit, Buffer A + Buffer B
CX-1 pH Gradient Starter Kit, Buffer A + Buffer B
Quantity:
125 mL
125 mL + 125 mL
250 mL
250 mL + 250 mL
500 mL
missing translation for 'unitSize'
125mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 15105634

missing translation for 'mfr': Thermo Scientific™ 083272

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Obtain fast and highly robust, reproducible HPLC gradients using Thermo Scientific™ CX-1 pH Gradient Buffers. Simple to optimize and easily automated, these buffers are applicable for HPLC separation of the majority of monoclonal antibody (mAb) biotherapeutics drugs. CX-1 pH Gradient Buffers are ready to use with existing HPLC columns without the need to formulate mobile phases. The CX-1 buffers save time in method development, facilitate method transfer to QA/QC for a wide range of mAb charge variants through a generic HPLC-based approach to mAb characterization, and do not require time-consuming mobile phase adjustments.

pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.

  • Robust, reproducible gradients
  • Predict biomolecule pH
  • Simple method optimization
  • Easily automated
  • Applicable to the majority of MAbs

Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pH and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.

TRUSTED_SUSTAINABILITY

Specifications

Description CX-1 pH Gradient High Resolution Kit, Buffer A + Buffer B
For Use With (Equipment) LC Columns and Systems
Type CX-1 pH Gradient High Resolution Kit
Includes MAbPac SCX-10 Column (5 μm, 4 x 250 mm)
pH 5.6 to 10.2
Quantity 125 mL + 125 mL
Product Line Thermo Scientific
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.