missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pH Gradient Buffer Concentrates

Product Code. 15155624
Click to view available options
Description:
CX-1 pH Gradient Buffer A
CX-1 pH Gradient Buffer B
CX-1 pH Gradient Buffer Kit, Buffer A + Buffer B
CX-1 pH Gradient High Resolution Kit, Buffer A + Buffer B
CX-1 pH Gradient High Throughput Kit, Buffer A + Buffer B
CX-1 pH Gradient Starter Kit, Buffer A + Buffer B
Quantity:
125 mL
125 mL + 125 mL
250 mL
250 mL + 250 mL
500 mL
Unit Size:
125mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 15155624

Brand: Thermo Scientific™ 085348

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Obtain fast and highly robust, reproducible HPLC gradients using Thermo Scientific™ CX-1 pH Gradient Buffers. Simple to optimize and easily automated, these buffers are applicable for HPLC separation of the majority of monoclonal antibody (mAb) biotherapeutics drugs. CX-1 pH Gradient Buffers are ready to use with existing HPLC columns without the need to formulate mobile phases. The CX-1 buffers save time in method development, facilitate method transfer to QA/QC for a wide range of mAb charge variants through a generic HPLC-based approach to mAb characterization, and do not require time-consuming mobile phase adjustments.

pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.

  • Robust, reproducible gradients
  • Predict biomolecule pH
  • Simple method optimization
  • Easily automated
  • Applicable to the majority of MAbs

Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pH and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.

TRUSTED_SUSTAINABILITY

Specifications

Description CX-1 pH Gradient Buffer B
For Use With (Equipment) LC Columns and Systems
Type CX-1 pH Gradient Buffer B
Volume (Metric) 250 mL
pH 10.2
Quantity 250 mL
Standard Concentration 10X
Product Line Thermo Scientific
compliance-icons
Nombre del producto
  • CX-1 pH Gradient Buffer B (pH 10.2)
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Otros peligros
  • MIXTURE LIST-Contains : sodium hydroxide, chloroform
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado