missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Indicator Paper Roll

Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.

38.70 €

Specifications

For Use With (Application) pH testing
Type pH Indicator Paper
View More Specs

Products 3
Product Code Brand pH Range Price Quantity & Availability  
Product Code Brand pH Range Price Quantity & Availability  
15940872
View Documents
Fisherbrand™
90207.204231.1
4 to 7
38.70 €
4 units
Please to purchase this item. Need a web account? Register with us today!
15950872
View Documents
Fisherbrand™
90210.204231.1
6.4 to 8
38.70 €
4 units
Please to purchase this item. Need a web account? Register with us today!
15960872
View Documents
Fisherbrand™
90212.204231.1
8 to 10
38.70 €
4 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
  • Allows very simple pH measurement
  • Proven standard for many applications
Specifications

Specifications

pH testing
pH Indicator Paper
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ pH Indicator Paper Roll

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.