missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Indicator Paper Rolls

Product Code. 15545679 Shop All Fisherbrand Products
Click to view available options
pH Range:
0 to 10
0 to 11
1 to 14
5.5 to 9
6.4 to 8
Format:
Refill
Roll
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 15545679

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides rapid and precise pH readings. Fisherbrand™ pH Indicator Paper Rolls includesa color scale printed on the cover for convenience.

  • Filter paper strips soaked with one or several coloured indicators of pH
  • Economical selection of indicator papers cover the entire range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows colour changes in steps of pH2

Specifications

pH Range 5.5 to 9
Format Roll
Includes Color scale printed on cover
Quantity 1 Each
Length (Metric) 5 m
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.