missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Indicator Paper Sticks

Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don't need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.

Brand:  Fisherbrand™ 92122

 View more versions of this product

Product Code. 10070552

  • 16.05 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don't need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.

85mm x 6mm sticks with different indicator papers sealed onto each strip. The pH is determined by comparison with a color chart supplied. The sticks are sufficiently long to avoid contact between the fingers and the test sample.
  • Provides outstanding measurements safely over the designated pH range
  • Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
  • Indicator is covalently bonded on the pads and does not bleed into the sample – can be left in sample for an indefinite time without contamination
  • Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
  • Available in a flat box so they fit into your pocket
Specifications

Specifications

ISO 13485 certified
Strip
100 Sticks
6 to 10
Colour comparison chart
pH Indicator Paper Stick
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.