missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH/Ion Meter SevenDirect SD50
GREENER_CHOICE

The SevenDirect SD50 is an accurate and flexible benchtop meter for the determination of pH, ORP and ion concentrations. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

Brand:  Mettler Toledo™ 30671552

 View more versions of this product

Product Code. 17590996

  • 1410.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 %
0.002
SevenDirect SD50, Meter Only
Color TFT
-2000 to 2000 mV
-30°C to +130°C
pH/Ion Meter
3 or 4 digits (auto-range)
0.1, 1
-22°F to +266°F
195 x 205 x 65 mm
850 g
0.1 mV
PC, Printer, USB Stick
Single-channel
IP54
0.1, 0.01, 0.001
2000 Standard Measurements
-2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm)
0.1, 1 mV
±0.1°C between 0°C and 100°C
0.1°C
1.9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.