missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ pH Meter SevenDirect SD20
Shop All Mettler Toledo Products

Click to view available options
Type:
SevenDirect SD20 HA Kit
SevenDirect SD20 Kit
SevenDirect SD20 LowVol Kit
SevenDirect SD20 Organic Kit
SevenDirect SD20 Pure H2O Kit
SevenDirect SD20 Solids Kit
SevenDirect SD20 Stirrer Kit
SevenDirect SD20 Viscous Kit
SevenDirect SD20, Meter Only
missing translation for 'unitSize'
Each
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
Specifications
| Includes | Instrument, power s upply, protective cover, Get Started guide, electrode holder EasyPlace, appropriate electrode, calibration solutions, EasyMix magnatic stirrer |
| Accuracy (Temperature) | ±0.1°C between 0°C and 100°C |
| Accuracy (mV) | 0.002 mV |
| Accuracy (pH) | 0.002 |
| Data Management Details | PC, Printer, USB Stick |
| Measurement Modes | pH with mV and Temperature |
| Calibration | Linear/Segmented |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Range (mV) | -2000 to 2000 mV |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction