Learn More
Mettler Toledo™ pH Meter SevenDirect SD20
The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
1120.00 € - 1845.00 €
Specifications
Accuracy (pH) | 0.002 |
---|---|
Accuracy (Temperature) | ±0.1°C between 0°C and 100°C |
Data Management Details | PC, Printer, USB Stick |
Resolution (pH) | 0.001, 0.01, 0.1 |
Range (mV) | -2000 to 2000 mV |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17570996
|
Mettler Toledo™
30671550 |
SevenDirect SD20, Meter Only |
1120.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17511006
|
Mettler Toledo™
30671554 |
SevenDirect SD20 Kit |
1565.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17531006
|
Mettler Toledo™
30671556 |
SevenDirect SD20 HA Kit |
1610.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17541006
|
Mettler Toledo™
30671557 |
SevenDirect SD20 Organic Kit |
1745.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17551006
|
Mettler Toledo™
30671558 |
SevenDirect SD20 Pure H2O Kit |
1765.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17561006
|
Mettler Toledo™
30671559 |
SevenDirect SD20 Solids Kit |
1695.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17571006
|
Mettler Toledo™
30671560 |
SevenDirect SD20 Viscous Kit |
1845.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17581006
|
Mettler Toledo™
30671561 |
SevenDirect SD20 LowVol Kit |
1795.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17521006
|
Mettler Toledo™
30671555 |
SevenDirect SD20 Stirrer Kit |
1785.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
0.002 | |
PC, Printer, USB Stick | |
-2000 to 2000 mV | |
-22°F to +266°F | |
TFT, 7 in. Color | |
IP54 | |
850 g |
±0.1°C between 0°C and 100°C | |
0.001, 0.01, 0.1 | |
-30°C to +130°C | |
0.1°C | |
2000 Standard Measurements | |
195 x 205 x 65 mm | |
1.9 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.