missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ pH Paper

Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.

13.05 €

Especificaciones

Certifications/Compliance ISO 9001
Format Roll
Quantity 1 Each
Type pH Paper
Ver más especificaciones

Productos 10
Código de producto Marca pH Range Graduations Precio Cantidad y disponibilidad  
Código de producto Marca pH Range Graduations Precio Cantidad y disponibilidad  
10797941
missing translation for 'viewDocuments'
Macherey-Nagel™
90204
1 to 14 1, 2, 3, 5, 6, 7, 8, 9, 10, 12, 14
13.05 €
5m
Please to purchase this item. Need a web account? Register with us today!
10098250
missing translation for 'viewDocuments'
Macherey-Nagel™
90205
0.5 to 5.5 0.5, 1.0, 1.5, 2.0, 2.5, 3.0, 3.5, 4.0, 4.5, 5.0, 5.5
13.05 €
Each
Please to purchase this item. Need a web account? Register with us today!
10464491
missing translation for 'viewDocuments'
Macherey-Nagel™
90201
1 to 11 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11
13.05 €
Each
Please to purchase this item. Need a web account? Register with us today!
10323361
missing translation for 'viewDocuments'
Macherey-Nagel™
90206
3.8 to 5.8 <3.8, 3.8, 4.1, 4.3, 4.5, 4.7, 4.9, 5.2, 5.5, 5.8 >5.8
13.05 €
5m
Please to purchase this item. Need a web account? Register with us today!
10539641
Ver productos accesorios missing translation for 'viewDocuments'
Macherey-Nagel™
90207
4 to 7 4.0, 4.3, 4.6, 4.9, 5.2, 5.5, 5.8, 6.1, 6.4, 6.7, 7.0
13.05 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
10549641
missing translation for 'viewDocuments'
Macherey-Nagel™
90208
5.4 to 7 <5.4, 5.4, 5.7, 6.0, 6.2, 6.4, 6.7, 7.0 >7.0
13.05 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
10559641
Ver productos accesorios missing translation for 'viewDocuments'
Macherey-Nagel™
90209
5.5 to 9 5.5, 6.0, 6.5, 7.0, 7.5, 8.0, 8.5, 9.0
13.05 €
5m
Please to purchase this item. Need a web account? Register with us today!
10570022
missing translation for 'viewDocuments'
Macherey-Nagel™
90210
6.4 to 8 <6.4, 6.4, 6.6, 6.8, 7.0, 7.2, 7.4, 7.6, 7.8, 8.0, >8.0
13.05 €
5m
Please to purchase this item. Need a web account? Register with us today!
10613141
missing translation for 'viewDocuments'
Macherey-Nagel™
90211
7.2 to 9.7 <7.2, 7.2, 7.5, 7.8, 8.1, 8.4, 8.7, 9.0, 9.3, 9.7, >9.7
13.05 €
5m
Please to purchase this item. Need a web account? Register with us today!
10527341
Ver productos accesorios missing translation for 'viewDocuments'
Macherey-Nagel™
90213
9 to 13 9.0, 9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0
13.05 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.

  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Especificaciones

Especificaciones

ISO 9001
Roll
1 Each
pH Paper
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado