Learn More
Fisherbrand™ pH/Pt1000 Electrode
This gel electrode with anti-shock epoxy body and protected head for the field is ideal for measuring 2 parameters pH and temperatures.
Brand:  Fisherbrand™ 5732528A4-003IU
 View more versions of this product
Product Code. 11729808
  • 115.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A single probe for the measurement of 2 parameters pH and temperature
  • Pt1000 sensor compatible with pH meters 
  • Gel electrode with anti-shock epoxy body and protected head for the field
  • Laboratory electrolyte glass electrode liquid
  • Supplied with 1 m cable BNC or DIN plug for pH and 2 banana plugs 4 mm for the temperature
Specifications
Specifications
DIN Plug
120 mm
0°C to 80°C
12 mm
0 to 14