Learn More
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper
Used for presumptive testing of suspicious stains. Phadebas™ Saliva Amylase-detection Forensic Paper is based on the Phadebas Amylase Test tablets but follows a separate method developed by the Forensic Biology labs.
193.00 € - 1145.00 €
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12895552
|
Magle Life Sciences™
1314 |
300 x 195 mm |
193.00 €
Pack of 12 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
12825562
|
Magle Life Sciences™
1317 |
395mm x 15m roll |
1145.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
Pre-manufactured papers;- Are applicable directly from package;
- Include Batch number and Expiry date;
- Are easy to handle and use;
- Have negligible batch variations;
- Give no difference between labs and consequently have higher credibility as evidence material