Learn More
Proteintech Phantom Dye Blue 516 Viability Dye
Phantom Dye Blue 516 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00007-100TESTS
Product Code. 17358252
  • 60.00 € / 100 tests
Explore more special offers
Description
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Blue 516 is a water soluble violet fluorescent dye excited by the 488 nm laser making it an excellent addition to your multicolor panel.
Specifications
Specifications
Flow Cytometry
516 Emission
100 Tests