Learn More
Proteintech Phantom Dye Red 710 Viability Dye
Phantom Dye Red 710 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00001-100TESTS
Product Code. 17398242
  • 60.00 € / 100 tests
Explore more special offers
Description
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Red 710 is a water soluble red fluorescent dye excited by red lasers (633-647 nm) making it an excellent addition to your multicolor panel.
Specifications
Specifications
Flow Cytometry
710 Emission
100 Tests