Learn More
Proteintech Phantom Dye Violet 450 Viability Dye
Phantom Dye Violet 450 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00004-100TESTS
Product Code. 17328252
  • 60.00 € / 100 tests
Explore more special offers
Description
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Violet 450 is a water soluble violet fluorescent dye excited by the Violet laser (405 nm), emitting at 450 nm, making it an excellent addition to your multicolor panel.
Specifications
Specifications
Flow Cytometry
450 Emission
100 Tests