missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Köttermann Pharmacy Fume Hood without Media
This Pharmacy Fume Hood without Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.
2735.00 € - 3025.00 €
Specifications
Dimensions (L x W x H) | 640 x 900 x 1350 mm |
---|---|
Voltage | 230 V |
Weight (Metric) | 115 kg |
Product Code | Brand | Configuration Options | Plug Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration Options | Plug Type | Price | Quantity & Availability | ||||
17352179
|
Köttermann
451-00015 |
No Ventilation Fan | Plug Type D (Old British Plug) |
2735.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17392179
|
Köttermann
451-00019 |
No Ventilation Fan | Schuko Plug |
2745.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17332189
|
Köttermann
451-00027 |
No Ventilation Fan | GB Plug |
2735.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17342179
|
Köttermann
451-00014 |
Ventilation Fan | Plug Type D (Old British Plug) |
3015.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17382179
|
Köttermann
451-00018 |
Ventilation Fan | Schuko Plug |
3025.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17322189
|
Köttermann
451-00026 |
Ventilation Fan | GB Plug |
3015.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For working safely with dust-forming preparations and volatile chemicals
- Long-lasting steel construction with one-piece polyacrylic sash
- Ergonomic features to improve workflow
- Low, compact design for small laboratories
Specifications
640 x 900 x 1350 mm | |
230 V | |
115 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title