Learn More
Labcold™ Pharmacy and Vaccine Refrigerator, Free Standing
Professional pharmacy refrigerator for the safe storage of cold chain medicines.
Brand:  Labcold™ RLDF1010A
Product Code. 15873651
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For the safe storage of cold chain medicines as advised by Public Health England in Protocol for Ordering, Storing and Handling Vaccines.
  • Fitted with an IntelliCold™ controller
  • Self-closing door
  • Door lock
  • Digital temperature control
  • Digital temperature display
  • Internal light
  • High/low temperature alarm
  • Min/max temperature recording
  • Automatic temperature and fridge function data recording
  • Door open alarm
  • Mains failure alarm
  • Alarm battery back-up
  • Off cycle auto defrost
  • Fan-assisted cooling
  • Remote alarm contacts
  • Access port
  • Low GWP refrigerant
  • DH Green Book Guidelines compliant
  • Five-year warranty
Specifications
Specifications
340 L
Solid
6
1500 x 600 x 700 mm
1.52 kWh/24 h
2°C to 8°C
Right
Powder-coated Steel
1358 x 480 x 500 mm
79 kg