missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PhastGel™ 8 to 25%, Tris-Acetate, 0.45 mm, Protein Gel

Separation Range: 50 to 30 Mr; Separation Type: Native

Brand:  Cytiva 17-0542-01


Product Code. 10309294

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing and native
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling

PhastGel homogenous gels

  • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
  • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution

PhastGel gradient gels

  • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
TRUSTED_SUSTAINABILITY
Specifications

Specifications

125°C
PhastGel Gradient – 8-25
0.45 mm
Gradient Page
50 to 30 Mr
Analytical Chemistry and Research
82° to 86°C
10/Pk.
4 - 8°C
50 mm
1.9 in.
45 mm
1.73 in.
Polyester
8 to 25%
Tris-Acetate
Protein
Native
GelBond polyester film
71.08
6% T/3% C
0.017 in.
4.4 cm
1.9 in.
5 cm
4.4 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B

Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Acrylamide
  • UNKNOWN ACUTE-85 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers