missing translation for 'onlineSavingsMsg'
Learn More
Phenamil, Tocris Bioscience™ Product Code.: 30001495

Phenamil, Tocris Bioscience™

Product Code. 30001495
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001495

Brand: Tocris Bioscience 3379/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TRPP3-mediated currents; also inhibits epithelial Na+ channels

TRPP3 channel inhibitor (IC50 = 0.14 μM). Also inhibits epithelial Na+ channels (Kd = 0.4 nM for a high affinity site on the epithelial Na+ channel). Derivative of amiloride (Cat. No. 0890).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1161-94-0
Molecular Formula C13H16ClN7O4S
Molecular Weight (g/mol) 401.826
InChI Key MHPIZTURFVSLTJ-UHFFFAOYSA-N
Synonym phenamil methanesulfonate, phenamil methanesulfonate salt, 3,5-diamino-6-chloro-n-imino phenylamino methyl pyrazinecarboxamide methanesulfonate salt, lopac-p-203, phenamil methanesulfonate salt, solid, phenamil methanesulfonate free base, 3,5-diamino-6-chloro-n-n-phenylcarbamimidoyl pyrazine-2-carboxamide methanesulfonate, 3,5-diamino-6-chloro-n-imino phenylamino methyl-pyrazinecarboxamide methansulfonateShow MoreShow Less
PubChem CID 9604979
IUPAC Name 3,5-diamino-N-[amino(anilino)methylidene]-6-chloropyrazine-2-carboxamide;methanesulfonic acid
SMILES CS(=O)(=O)O.C1=CC=C(C=C1)NC(=NC(=O)C2=C(N=C(C(=N2)Cl)N)N)N

Specifications

Quantity 10 mg
Formula Weight 401.83
Percent Purity >99%
Chemical Name or Material Phenamil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenamil, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.