missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl alcohol, 99%, Thermo Scientific Chemicals

23.62 € - 401.00 €

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
SMILES OCCC1=CC=CC=C1
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10133230
View Documents Promotion Details
Thermo Scientific Acros
130180050
5 mL Glass bottle
23.62 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10246563
View Documents Promotion Details
Thermo Scientific Acros
130181000
100 mL Glass bottle
25.02 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10521784
View Documents Promotion Details
Thermo Scientific Acros
130182500
250 mL Glass bottle
32.86 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10123230
View Documents Promotion Details
Thermo Scientific Acros
130180010
1 L Glass bottle
66.13 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10562141
View Documents Promotion Details
Thermo Scientific Acros
130180025
2.5 L Glass bottle
149.75 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10792754
View Documents Promotion Details
Thermo Scientific Acros
130180100
10 L Plastic Drum
401.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-12-8
122.17
WRMNZCZEMHIOCP-UHFFFAOYSA-N
6054
OCCC1=CC=CC=C1
C8H10O
MFCD00002886
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
CHEBI:49000

Specifications

60-12-8
Colorless
219.0°C
Authentic
C8H10O
1.5300 to 1.5350
06, 478
1.02
Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
OCCC1=CC=CC=C1
122.17
CHEBI:49000
122.17
Liquid
-27.0°C
1.0200g/mL
102°C
98.5% min. (GC)
C6H5CH2CH2OH
MFCD00002886
15, 7338
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
WRMNZCZEMHIOCP-UHFFFAOYSA-N
2-phenylethanol
6054
7.58 mPa.s (25°C)
99%
Phenethyl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-456-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only