missing translation for 'onlineSavingsMsg'
Learn More

Phenol, 99.5%, unstab., Thermo Scientific Chemicals

Product Code. 15491997
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

As a precursor to many materials and useful compounds. Its major uses involve its conversion to plastics or related materials. Phenol and its chemical derivatives are key for building polycarbonates, epoxies, Bakelite, nylon, detergents, and numerous pharmaceutical drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-95-2
Melting Point 40°C to 43°C
Density 1.071 g/mL
Boiling Point 181°C
Flash Point 79°C (175°F)
Odor Sweet
Molecular Formula C6H6O
MDL Number MFCD00002143
Quantity 100 g
UN Number UN1671
Beilstein 969616
Sensitivity Hygroscopic
Merck Index 14,7241
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
Solubility Information Moderately soluble in water (8.4% @ 20°C),alcohol,ether,chloroform,ethyl acetate,toluene,and glycerol.
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 996
ChEBI CHEBI:15882
Formula Weight 94.11
Percent Purity 99.5%
Physical Form Solid Melt
Chemical Name or Material Phenol, UnStabilized
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis