missing translation for 'onlineSavingsMsg'
Learn More

Phenol, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10479803
Click to view available options
Quantity:
50 g
1 kg
2.5 kg
5 kg
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39.0°C to 42.0°C
Color Colorless
Density 1.0700g/mL
Boiling Point 182.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula C6H5OH
Quantity 5 kg
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Specific Gravity 1.07
Solubility Information Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
Viscosity 3.437 mPa.s (50°C)
Formula Weight 94.11
Vapor Pressure 0.4mbar at 20°C
Percent Purity 99%
Grade Extra Pure
Physical Form Crystals or Fused Mass
Chemical Name or Material Phenol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'